Hach HQ2100 Portable Multiparameter with Dissolved Oxygen Electrode, 1 m Cable

Representative image only.
Hach Mfr # LEV015.53.21004 – Item # EW-59005-14
A robust and intuitive multiparameter meter
  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 10,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
  • 1 m dissolved oxygen electrode
  • 1 year warranty

PRODUCT OPTIONS

$2,710.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypepH, conductivity, TDS, salinity, DO, ORP
  • Display type536x336 Mono-TFT
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.001/0.01/0.1
  • pH AccuracyProbe specific
  • Buffer RecognitionIUPAC, USA, DIN, Custom
  • CalibrationUp to 3 points
  • Min mV (mV)-1200
  • mV Resolution0.1 mV
  • mV AccuracyProbe specific
  • Min Conductivity0.01 µS/cm
  • Max Conductivity200 mS/cm
  • Conductivity Resolution0.01 µS/cm to 0.1 mS/cm upon selected measuring range
  • Conductivity AccuracyProbe specific
  • Min TDS0.01 g/L
  • Max TDS50 g/L
  • TDS Resolution0.01 mg/L up - 0.1 g/L (range dependent)
  • TDS AccuracyProbe specific
  • Min Salinity42 ppt or %
  • Max Salinity0 ppt or %
  • Salinity Resolution0.01 (range dependent)
  • Salinity AccuracyProbe specific
  • Min Dissolved Oxygen0.0 mg/L
  • Max Dissolved Oxygen23 mg/L
  • Dissolved Oxygen Resolution0.1 mg/L
  • Dissolved Oxygen Accuracy±0.1 mg/L from 0 to 8 mg/L, or ±0.2 mg/L for greater than 8 mg/L
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature Resolution 0.1°C
  • Temperature Accuracy±0.3 °C (±0.54 °F)
  • Temperature compensationATC
  • OutputUSB
  • Data Logging (points)Yes
  • Memory Size10,000 data points
  • WaterproofYes, IP67
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Battery1 rechargeable lithium-ion (included)
  • Length (cm)22
  • Width (cm)9.7
  • Height (cm)6.3
  • Width (in)3 13/16
  • Length (in)8 21/32
  • Height (in)2 31/64
  • Number of Positions1
  • NIST Calibration IncludedNo
  • Probe Included1 m cable DO electrode
  • DescriptionPortable Multi-Meter with Dissolved Oxygen Electrode, 1 m Cable
  • Warranty1 year
1 Year WarrantyConformite EuropeenneMade in the U.S.A.

Includes

Meter, LDO101 DO electrode (59004-57), rechargeable battery, USB power adapter, and USB cable.

Key Features

  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 10,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
  • 1 m dissolved oxygen electrode
  • 1 year warranty

More About this Item

Hach portable HQ meters offer a versatile range of measurements, including pH, conductivity, total dissolved solids (TDS), salinity, dissolved oxygen (DO), oxidation reaction potential (ORP), and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.

The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat