Hach HQd11d Portable Starter Package with MTC101 ORP Electrode

Representative image only.
Hach Mfr # 8506500 – Item # EW-59004-14
IntelliCA™ smart probes have ''plug-and-play'' technology for maximum flexibility
  • HQd meters with IntelliCAL probes packaged together for your convenience
  • Option to select from 13-lanuages in setup mode
  • Store up to 500 GLP-compliant data sets
  • Rugged, IP67-rated waterproof housing
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeGraphic LCD with backlight
  • Min pH (pH)2
  • Max pH (pH)14
  • pH Resolution0.001
  • pH Accuracy±0.02
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 3 points
  • Min mV (mV)-1200
  • Max mV (mV)1200
  • mV Resolution0.1 mV
  • mV Accuracy±0.02
  • Temperature compensationAutomatic or manual
  • InterfaceUSB
  • WaterproofYes
  • Width (cm)17.4625
  • Height (cm)23.65375
  • Width (in)6 7/8
  • Length (in)3 3/8
  • Height (in)9 5/16
  • DescriptionPortable Starter Package with MTC101 ORP Electrode
  • Warranty1 year
1 Year WarrantyIP 67

Key Features

  • HQd meters with IntelliCAL probes packaged together for your convenience
  • Option to select from 13-lanuages in setup mode
  • Store up to 500 GLP-compliant data sets
  • Rugged, IP67-rated waterproof housing
  • 1 year warranty

More About this Item

Versatile HQd meters offer a large backlit display with an intuitive user-interface. The meter prompts you through calibration, setup, and measurements. Automatic or manual temperature compensation, auto buffer recognition, and auto calibration for up to three points with USA, NIST, DIN, and custom buffers.

Meter can automatically or manually log up to 500 measurements with probe serial number, current calibration data, user ID, sample ID, time, and date. Use the USB port to download data to a PC or flash drive.

The HQd series uses precalibrated Hach IntelliCAL smart probes that store their own calibration data and identification. The meter recognizes each probe, so you can easily move between meters without recalibrating. Kit includes standard laboratory probe. Order optional rugged probe for field applications; all connections between the meter and probe are secure and waterproof.


What's Included: Hach HQ11d portable meter, ORP laboratory probe with 3.3-ft (1-m) cable (59004-66), meter stand, electrode stand, and four AA batteries.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat