Hach HS-C HYDROGEN SULFIDE TEST KIT
Representative image only.
Hach Mfr # 2537800 – Item # EW-05545-99
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year Warranty
Includes
Reaction vial, effervescent tablets, test papers, comparator chart, and instructions.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
  • 1 year warranty
More About this Item
The hydrogen sulfide test kit is an economical method for determining hydrogen sulfide content in water samples. Results are obtained by comparison of the color to a printed chart.