Hach IRON STRIPS Total Dissolved Iron Test Strips, 25
Representative image only.
Hach Mfr # 2745325 – Item # EW-05546-32
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$34.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestIron, total dissolved
  • Measurement Points0, 0.15, 0.3, 0.6, 1, 2, 5 ppm
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionTotal Dissolved Iron Test Strips, 25
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test total dissolved Iron as Fe from 0 to 5 mg/L. 25 test strips included.