Hach LZW5060.97.0002 SensIon+ Conductivity Cell, General
Representative image only.
Hach Mfr # LZW5060.97.0002 – Item # EW-19604-51
Conductivity cells for general or robust applications
  • Two measuring ranges
  • Range of 0.2 µS/cm to 200 mS/cm
  • Plastic body with Platinum sensor
  • Replacement cell for 19604-01 clean water kit.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSensIon+ Conductivity Cell, General
  • Warranty1 year
1 Year Warranty
Key Features
  • Two measuring ranges
  • Range of 0.2 µS/cm to 200 mS/cm
  • Plastic body with Platinum sensor
  • Replacement cell for 19604-01 clean water kit.
  • 1 year warranty
More About this Item
General cell measures from 0.2 µS/cm to 200 mS/cm. Robust cell measures from 5 µS/cm to 50 mS/cm.