Hach NITRATE STRIPS Nitrate and Nitrite Test Strips, 25
Representative image only.
Hach Mfr # 2745425 – Item # UX-05546-36
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$31.00USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • TestNitrate and nitrite
  • Measurement Points0 to 50 ppm; 0 to 3 ppm
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionNitrate and Nitrite Test Strips, 25
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Tests Nitrate from o to 50 mg/L and Nitrite from 0 to 3 mg/L. 25 test strips included.