Hach sensION 3 Benchtop pH/mV Meter, 110 VAC

Representative image only.
Hach Mfr # 51750-10 – Item # EW-59004-20
Antiglare coating for easy viewing
  • Unique “All-in-One” five-pin connector for simple setup
  • Stable readings in seconds!
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)19.99
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232
  • Power (VAC)115
  • Power (Hz)60
  • Width (cm)15.24
  • Height (cm)26.035
  • Width (in)6
  • Length (in)3 1/2
  • Height (in)10 1/4
  • DescriptionBenchtop pH/mV Meter, 110 VAC
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67

Key Features

  • Unique “All-in-One” five-pin connector for simple setup
  • Stable readings in seconds!
  • 3 year warranty

More About this Item

sensION 3 Meter measures pH, mV, and temperature. Other features include built-in communications protocol for downloading and printing data and a user-activated calibration review that shows the date and time of last calibration, slope, and buffers used.

Use these meters for your most demanding applications. Meters feature a chemical-resistant, dust-proof, and water-resistant, IP67 case—perfect for rugged environments. Automatic temperature compensation and automatic buffer recognition provide for accurate readings.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat