Hach TNT822 TNTplus® Spectrophotometer Chemistry, High Range COD, 20 to 1500 mg/L; 25/PK
Representative image only.
Hach Mfr # TNT822 – Item # EW-99515-17
Perform common methods with these pre-made reagent tests
  • Measures: High Range COD, 20 to 1500 mg/L Package: 25
PRODUCT OPTIONS
$60.99USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHigh Range COD, 20 to 1500 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, High Range COD, 20 to 1500 mg/L; 25/PK
Key Features
  • Measures: High Range COD, 20 to 1500 mg/L Package: 25
More About this Item
Dozens of options to cover most applications!