Hach TNT843 TNTplus® Spectrophotometer Chemistry, Low Range Phosphorus, 0.05 to 1.5 mg/L PO4-P; 25/PK
Representative image only.
Hach Mfr # TNT843 – Item # EW-99515-34
Perform common methods with these pre-made reagent tests
  • Measures: Low Range Phosphorus, 0.05 to 1.5 mg/L PO4-P
PRODUCT OPTIONS
$64.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLow Range Phosphorus, 0.05 to 1.5 mg/L PO4-P
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Low Range Phosphorus, 0.05 to 1.5 mg/L PO4-P; 25/PK
Key Features
  • Measures: Low Range Phosphorus, 0.05 to 1.5 mg/L PO4-P
More About this Item
Dozens of options to cover most applications!