Hach TNT872 TNTplus® Spectrophotometer Chemistry, Volatile Acids, 50 to 2500 mg/L; 25/PK
Representative image only.
Hach Mfr # TNT872 – Item # EW-99515-51
Perform common methods with these pre-made reagent tests
  • Measures: Volatile Acids, 50 to 2500 mg/L
PRODUCT OPTIONS
$116.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVolatile Acids, 50 to 2500 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Volatile Acids, 50 to 2500 mg/L; 25/PK
Key Features
  • Measures: Volatile Acids, 50 to 2500 mg/L
More About this Item
Dozens of options to cover most applications!