Hamilton 79427 PRP-1 Reverse Phase Column, Analytical, 250x4.1mm ID; 10um
Representative image only.
Hamilton Mfr # 79427 – Item # EW-01941-04
High capacity and rapid flow rates for fast analysis
  • Compatible with any concentration of organic solvent
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePolystyrenedivinylbenzene
  • Column Particle Size (µm)10
  • DescriptionPRP-1 Reverse Phase Column, Analytical, 250x4.1mm ID; 10um
  • Warranty18 months
18 Month Warranty
Key Features
  • Compatible with any concentration of organic solvent
  • 18 month warranty
More About this Item
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene)stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-1 Reverse Phase Columns contain poly(styrenedivinylbenzene). Large surface area (415 m2/g) allows high capacity and rapid flow rates for fast analysis. Ideal for separating strong acids and bases. Efficiency of 30,000 to 50,000 plates/meter.