Hamilton 79442 PRP-X200 Cation Exchange Column, Analytical, 250x4.1mm ID; 10um
Representative image only.
Hamilton Mfr # 79442 – Item # EW-01943-02
Efficient separations with durable media
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePolystyrenedivinylbenzene
  • Column Particle Size (µm)10
  • DescriptionPRP-X200 Cation Exchange Column, Analytical, 250x4.1mm ID; 10um
  • Warranty18 months
18 Month Warranty
Key Features
  • 18 month warranty
More About this Item
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene)
stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-X200 Strong Cation Exchange Columns contain poly (styrene-divinylbenzene) sulfonate. These columns also offer superior, rapid resolution; exchange capacity is 0.035 mEq/g.