Hamilton 90126 CTFE-hubbed hypodermic needles, 26 gauge
Representative image only.
Hamilton Mfr # 90126 – Item # EW-07938-96
Provides accurate volume delivery
PRODUCT OPTIONS
$120.00USD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeLuer Tip
  • Needle Tip StyleNoncoring deflected
  • Needle Tip Number2
  • Needle Gauge26
  • Needle Length (in)2
  • DescriptionCTFE-hubbed hypodermic needles, 26 gauge
More About this Item
The syringe minimizes the gas exchange in analysis. The plunger is unsurpassed, while the tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.