Hand-Motion Shaker, 0 to 45° angle, 60 to 180 rpm, 115 VAC

Representative image only.
Mfr # SHK-COCK, 115V – Item # EW-04636-60
Ideal for DNA extractions, pretreatment of enzyme reactions, treatment of reagents, and gel treatment for electrophoresis
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max Speed (rpm)180
  • Width (in)5.75
  • Height (in)6
  • Depth (in)5.5
  • Width (cm)14.605
  • Height (cm)15.24
  • Depth (cm)13.97
  • Power (VAC)115
  • Power (Hz)50/60
  • DescriptionHand-Motion Shaker, 0 to 45° angle, 60 to 180 rpm, 115 VAC
  • Warranty1 year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Perform continuous or timed treatments using built-in timer. Adjust shaker platform for a more vertical or more horizontal shaking angle, giving you ability to customize for your specific applications. Two adhesive rubber pads are located at the base. Shaker requires one of four platforms sold separately.

What's included: a 5-ft, two-wire cord & plug

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat