Hardman 13230A/B-SY High-Peel Strength Epoxy Syringe, 30 mL; 1/Pack
Representative image only.
Hardman Mfr # 13230A/B-SY – Item # EW-08778-50
Apply amount necessary, then reseal syringe for later use
  • Quick-dispensing provides convenient repair work
PRODUCT OPTIONS
$22.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Curing time (days)7
  • Curing time (hrs)96
  • Set time (mins)240
  • Set time (hrs)4
  • Min operating temperature (° F)-40
  • Max operating temperature (° F)250
  • DescriptionHigh-Peel Strength Epoxy Syringe, 30 mL; 1/Pack
Includes
Syringe cap and ten mixing sticks.
Key Features
  • Quick-dispensing provides convenient repair work
More About this Item
Double-barrel syringe epoxies provide the appropriate amount of two-component epoxies for the job at hand. There is no need to worry whether or not the correct amount is dispensed. Just dispense, mix and apply to the surface being repaired.