Hardman Double Bubble® Epoxy Pouches, Non-Sag Extra Fast Setting, 3.5g; 10/Pk
Representative image only.
Hardman Mfr # 4008-BG10 – Item # EW-08778-04
Complete selection of epoxies and urethane for all your maintenance needs
  • Single-use adhesives are perfect for quick repair jobs
  • Extra fast setting, non-sag (thixotropic) adhesive for quick repairs
  • Bonds to wood, glass, metal, stone, ceramics and leather
PRODUCT OPTIONS
$71.00USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$71.00USD / Pkg of 10
  • 10+$63.90USD / Pkg of 10
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Curing time (days)1
  • Curing time (mins)1440
  • Curing time (hrs)24
  • Set time (mins)5
  • Min operating temperature (° F)-20
  • Max operating temperature (° F)180
  • Number of Pieces per Set10
  • DescriptionEpoxy Pouches, Non-Sag Extra Fast Setting, 3.5g; 10/Pk
Includes
10 pouches and mixing sticks.
Key Features
  • Single-use adhesives are perfect for quick repair jobs
  • Extra fast setting, non-sag (thixotropic) adhesive for quick repairs
  • Bonds to wood, glass, metal, stone, ceramics and leather
More About this Item
Epoxies and urethanes come in handy 3.5-gram, two-component packets with mixing sticks. Choose from individual packets or two kit options providing an assortment of epoxies and urethanes.