Hardman 4024-BG10 Very Flexible Urethane Pouches, 3.5g; 10/Pack
Representative image only.
Hardman Mfr # 4024-BG10 – Item # EW-08778-20
Complete selection of epoxies for all your maintenance needs
  • Single-use adhesives are perfect for quick repair jobs
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Curing time (days)7
  • Curing time (hrs)96
  • Set time (mins)8
  • Min operating temperature (° F)-40
  • Max operating temperature (° F)150
  • Number of Pieces per Set10
  • DescriptionVery Flexible Urethane Pouches, 3.5g; 10/Pack
Key Features
  • Single-use adhesives are perfect for quick repair jobs
More About this Item
Epoxies and Urethanes come in handy 3.5-gram, two-component packets and mixing sticks. Choose from individual packets or two kit options providing an assortment of 100 epoxies and urethanes. Kits are available as a tool box with a removable tray or a satchel case divided into 15 compartments.
What's included: packets include 10 epoxy or urethane packs and mixing sticks. Kits include 100 mixing sticks; 30 packets of quick-set epoxy (08778-02); 10 packets each of quick-set, non-sag epoxy (08778-04) and general-purpose epoxy (08778-06); five packets each of high peel-strength epoxy (08778-08), machinable epoxy (08778-10), wet-surface epoxy (08778-12), general-purpose urethane (08778-20), high peel-strength urethane (08778-22), high shear-strength urethane (08778-24), regular setting epoxy, water clear epoxy, fast setting silicone, and hand cleaner.