Hardman EMIX-100SAT-CP Epoxy and Urethane Adhesive Satchel Case Kit Assortment; 200/Kit
Representative image only.
Hardman Mfr # EMIX-100SAT-CP – Item # EW-08778-32
Complete selection of epoxies for all your maintenance needs
  • Single-use adhesives are perfect for quick repair jobs
PRODUCT OPTIONS
$334.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Pieces per Set200
  • DescriptionEpoxy and Urethane Adhesive Satchel Case Kit Assortment; 200/Kit
Key Features
  • Single-use adhesives are perfect for quick repair jobs
More About this Item
Epoxies and Urethanes come in handy 3.5-gram, two-component packets and mixing sticks. Choose from individual packets or two kit options providing an assortment of 100 epoxies and urethanes. Kits are available as a tool box with a removable tray or a satchel case divided into 15 compartments.
What's included: packets include 10 epoxy or urethane packs and mixing sticks. Kits include 100 mixing sticks; 30 packets of quick-set epoxy (08778-02); 10 packets each of quick-set, non-sag epoxy (08778-04) and general-purpose epoxy (08778-06); five packets each of high peel-strength epoxy (08778-08), machinable epoxy (08778-10), wet-surface epoxy (08778-12), general-purpose urethane (08778-20), high peel-strength urethane (08778-22), high shear-strength urethane (08778-24), regular setting epoxy, water clear epoxy, fast setting silicone, and hand cleaner.