Hardy Diagnostics Butterfield's phosphate buffer, prepared; 500 mL, 10/pk
Representative image only.
Hardy Diagnostics Mfr # U190 – Item # EW-14204-56
Save time and money with ready-made bottles
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the preparation of dilutions and other laboratory tests
PRODUCT OPTIONS
$175.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Phosphate Buffer (Butterfield's), pH 7.2
  • Capacity (mL)500
  • Capacity (L)0.5
  • DescriptionButterfield's phosphate buffer, prepared; 500 mL, 10/pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the preparation of dilutions and other laboratory tests
More About this Item
Bottles are tested and standardized to ensure consistent and reliable performance. Each bottle includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Bottle and pack sizes vary.