Harvard Apparatus 53-2022W Frame Clamps, Nylon and SS, open-sided, 5/Pk
Representative image only.
Harvard Apparatus Mfr # 53-2022W(X5 PIECES) – Item # EW-08024-28
Glass fiber-reinforced nylon and stainless steel components provide enormous strength
  • This material has a melting point of 221°C.
  • Resistance to salt solutions and solvents.
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialFiberglass-reinforced Nylon and Stainless Steel
  • Clamp TypeFrame and Rod
  • Accepts Rod Diameter (in)0.5
  • DescriptionFrame Clamps, Nylon and Ss, Open-Sided, 5/Pk
Key Features
  • This material has a melting point of 221°C.
  • Resistance to salt solutions and solvents.
More About this Item
Use with rods, equipment clamps, or adapters to position your equipment exactly as you want. Thumbscrews are included.