Hayward DAB1005UEE Diaphragm Valve, PVC, 1/2" True Union Connection
Representative image only.
Hayward Mfr # DAB1005UEE – Item # EW-07390-47
Designed for use in corrosive environments
  • Multi-turn handle gives you precise control
  • Position indicator shows exact location of valve
  • Self draining allows for little dead volume in valve
  • PVC body and EPDM diaphragm
  • Tru-union connections
PRODUCT OPTIONS
$118.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/2" true union socket
  • Connection TypeTrue union socket
  • Inlet Port1/2" true union socket
  • Cv3.5
  • Wetted MaterialsPVC
  • Working Pressure (PSI)225
  • DescriptionDiaphragm Valve, PVC, 1/2" True Union Connection
Key Features
  • Multi-turn handle gives you precise control
  • Position indicator shows exact location of valve
  • Self draining allows for little dead volume in valve
  • PVC body and EPDM diaphragm
  • Tru-union connections