Hayward F3AB00020E Single-Length Filtration Bags; 25 µm, 50/Pk
Representative image only.
Hayward Mfr # PO025G1PRSH – Item # EW-29990-58
Conveniently collect all filtered particles within the bag
  • Bags are constructed of glass-reinforced polypropylene (PP)
PRODUCT OPTIONS
$386.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene, glass reinforced
  • Micron Rating25
  • Descriptionsingle-length bags; 25 µm, 50/pack
Key Features
  • Bags are constructed of glass-reinforced polypropylene (PP)
More About this Item
Hayward(R) bags are made from nonwoven polypropylene felt. The single-length bags have a surface area of 2.0 ft2; double length bags have a surface area of 4.1 ft2. Bags are available in mulitple pore sizes.