Hayward FV10050SA Foot valve screen for ball valve 01341-00, 1/2"
Representative image only.
Hayward Mfr # FV10050SA – Item # EW-01341-50
Unique square-cut elastomer seat provides seal at very low back pressures
PRODUCT OPTIONS
$68.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Cv4.6
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC, EPDM
  • Working Pressure (PSI)150
  • DescriptionFoot valve screen for ball valve 01341-00, 1/2"
More About this Item
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.
The foot valve screen helps prevent unwanted material from entering the system.