Hayward HCTB1200STACTV-EAU29 Electrically Actuated PVC True Union Ball Valve, 2", 115 VAC
Representative image only.
Hayward Mfr # HCTB1200STACTV-EAU29 – Item # EW-01347-10
  • Ideal for remote control of process fluids
  • Precision machined stems specifically designed for actuated valves
  • Adjustable and reversible PTFE(R) PTFE seats
  • One-piece PVC or CPVC body construction
  • Includes two Viton(R) seals for double leakproof protection
PRODUCT OPTIONS
$625.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))2
  • Outlet Port2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port2" NPT(F)
  • Cv140
  • Max Pressure (PSI)225
  • Power (VAC)115
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC, Viton
  • DescriptionElectrically Actuated PVC True Union Ball Valve, 2", 115 VAC
Key Features
  • Ideal for remote control of process fluids
  • Precision machined stems specifically designed for actuated valves
  • Adjustable and reversible PTFE(R) PTFE seats
  • One-piece PVC or CPVC body construction
  • Includes two Viton(R) seals for double leakproof protection
  • Remote control capabilities via an auxiliary limit switch
  • Thermal overload circuit protects the motor from burnout and overheating
  • Easy connections via a wiring terminal strip
  • Unique mounting feature allows for easy conversion to manual operation
  • Mechanical brake for precise cycle control
  • Actuator comes mounted on the valve
  • NEMA 4X polypropylene housing stands up to harsh operating conditions