Hayward PVDF Needle Valve, 1/2" NPT(F), with PTFE Seat and FPM Seals
Representative image only.
Hayward Mfr # NVA5050T – Item # EW-03245-90
  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
PRODUCT OPTIONS
$156.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type1/2" NPT(F)
  • Cv0.79
  • Housing MaterialPVDF
  • Max Pressure (PSI)150
  • Max Pressure (bar)10.3
  • Max Temperature (° F)240
  • Max Temperature (° C)116
  • DescriptionPVDF Needle Valve, 1/2" NPT(F), with PTFE Seat and FPM Seals
Key Features
  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
More About this Item
Precision needle valves are available in four materials: PVC, CPVC, PP and PVDF. All feature an integrated stem with PTFE seat; fine pitch stem threads ensure precise adjustment. Valve body also includes flanges for panel mounting. Connections are standard pipe thread.