Hayward TC10100STE True Union Ball Check Valve, 1" socket and NPT(F) connectors
Representative image only.
Hayward Mfr # TC10100STE – Item # EW-01341-04
Unique square-cut elastomer seat provides seal at very low back pressures
  • Constructed of a PVC body and EPDM seal
PRODUCT OPTIONS
$104.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))1
  • Outlet Port1" NPT(F) and socket
  • Connection TypeNPT(F) and Socket
  • Inlet Port1" NPT(F) and socket
  • Cv28
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC, EPDM
  • Working Pressure (PSI)150
  • DescriptionTrue Union Ball Check Valve, 1" socket and NPT(F) connectors
Key Features
  • Constructed of a PVC body and EPDM seal
More About this Item
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.