HD Electric 9391 Digital High-Voltage TRMS Fork Head Ammeter
Representative image only.
HD Electric Mfr # 9391 – Item # EW-25751-03
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (amps)2000
  • DescriptionDigital High-Voltage TRMS Fork Head Ammeter
More About this Item
Ideally suited for taking spot-load studies at transformers or at any mid-span location, as well as phase balance studies on primary or secondary lines. This is a true RMS reading instrument. Its load-latching circuitry continually latches and locks on to peak system currents. It will display and hold the reading until the display is reset.