HD Electric DVM-I Single Stick Voltmeter, 0-20 kV, 31" Length
Representative image only.
HD Electric Mfr # DVM-I – Item # EW-20022-01
Get safe and accurate voltage readings
  • Feature a backlit display for low-light conditions, auto shut-off, and low-battery indicator
  • Powered by 9 V batteries
  • True RMS voltage measurements
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)0
  • DC Max (Volts)20000
  • AC Min (Volts)0
  • AC Max (Volts)20000
  • DescriptionSingle Stick Voltmeter, 0-20 kV, 31\" Length
  • Warranty1 Year
1 Year Warranty
Key Features
  • Feature a backlit display for low-light conditions, auto shut-off, and low-battery indicator
  • Powered by 9 V batteries
  • True RMS voltage measurements
  • 1 year warranty
More About this Item
The DigiVolt® series of digital voltmeters/phasing sets are designed to take voltage measurements and determine if two conductors are in-phase or out-of-phase on system voltages.
The DVM-I is a single-stick voltmeter with a ground lead designed for taking line-to-ground measurements up to 20 kV, and features peak hold mode for displaying the maximum voltage read until the display is manually reset.