Heavy-Duty Tubing Pinch Clamp, Polysulfone
Representative image only.
Item # EW-20001-00
Durable design handles higher working pressures
  • Quick installation with positive swing-away locking screw
  • Smooth contours allow for easy cleaning
$46.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolysulfone
  • DescriptionHeavy-Duty Tubing Pinch Clamp, Polysulfone
Key Features
  • Quick installation with positive swing-away locking screw
  • Smooth contours allow for easy cleaning
More About this Item
Use this clamp with tubing or hose - it will meet the working pressure of either. High-strength polysulfone closure provides excellent durability, can be continually reused, and resists most chemicals. Simply align the hose within the clamp; mating surfaces on the clamp indicate complete closure. Typical applications include sterile filling, media bag connections, pumping, liquid control and high-purity fluid transfers. FDA-compliant polysulfone material can be sterilized by autoclave or gamma radiation.