Hedland H601A-005 Flow Meter, 1/2in. Alum, 0.5/5.0 GPM Oil 1/2" Aluminum Body, NPTF Connection
Representative image only.
Hedland Mfr # H601A-005 – Item # EW-32508-01
Install in any position—360° rotatable guard/scale
  • Easier to read linear scale
PRODUCT OPTIONS
$266.10USD / Each
$310.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Operating Temperature (° F)240
  • Max Operating Temperature (° C)115.5
  • DescriptionFlow Meter, 1/2in. Alum, 0.5/5.0 GPM Oil 1/2" Aluminum Body, NPTF Connection
Key Features
  • Easier to read linear scale
More About this Item
FEATURES
  • Direct reading
  • Install in any posiiton
  • 360 degrees rotatable guard / scale
  • Easier to read linear scale
  • No flow straighteners or special piping required
  • Relatively insensitive to shock and vibration
  • Good viscosity stability
  • Temperature up to 240F
  • Accuracy ± 2% full scale
  • Repeatability ± 1%
  • Special scales available