Hedland HB110-375-PA Low-Power Turbine Flow Sensor, 0.3 to 3 GPM

Representative image only.
Hedland Mfr # B110-375-PA – Item # EW-32249-00
Display versions designed for use in hazardous process environments
Battery-powered displays - no wiring required and intrinsically safe
Large display face is easily readable from long distances
NEMA 4X enclosure is ideal for wet process environments

PRODUCT OPTIONS

$911.00USD / Each

$310.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±1% of reading
  • Repeatability±0.1%
  • Max Flow Rate (GPM)3
  • Max Operating Temperature (° F)325
  • Max Operating Temperature (° C)163
  • Pressure Drop At Maximum Flow (PSID)1.2
  • Output (Voltage)Yes
  • OutputSquare wave pulse
  • Display TypeNone
  • DescriptionLow-Power Turbine Flow Sensor, 0.3 to 3 GPM

More About this Item

The sensors feature 316 stainless steel (SS) body with 416 SS rotor and abrasion-resistant tungsten carbide bearings and shaft to endure corrosive water or other aggressive fluids. All sensors include flow straighteners for stabilizing turbulent flow allowing sensors to be mounted in any position*.
Use sensors with an existing monitor located locally or remotely. Sensors feature a pre-amplifier that must be powered and provides an output signal at distances up to 1000 feet. To interface the flow sensor, order multi-pin cable 32249-95.
Flowmeters include a flow sensor with a mounted or remote display. The display is a NEMA 4X (IP66) enclosure. For the remote system, the sensor may be located up to 10 feet away from the display.
* For highest accuracy, at least ten pipe diameters long on inlet and five on outlet.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat