HF Scientific 28336 Solid Validation Turbidity Standard, Mid NTU
Representative image only.
HF Scientific Mfr # 28336 – Item # EW-99900-54
Calibrate turbidimeter to ensure accurate readings
  • Typically around 7 NTU
  • Complies with US EPA Method 180.1
PRODUCT OPTIONS
$518.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHF Scientific 28336 Solid Validation Turbidity Standard, Mid NTU
Key Features
  • Typically around 7 NTU
  • Complies with US EPA Method 180.1