HF Scientific 52070 Hach ProCal Primary Standard for Process Turbidimeter 1720, 1.0 NTU
Representative image only.
HF Scientific Mfr # 52070 – Item # EW-99900-35
Instrument specific kits and standards ensure maximum accuracy
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
PRODUCT OPTIONS
$191.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • NTU Value1.0
  • Volume (mL)1000
  • Volume (oz)33.8
  • Use withHach 1720 Process Turbidimeters
  • DescriptionHach ProCal Primary Standard for Process Turbidimeter 1720, 1.0 NTU
  • Warranty1 year
1 Year Warranty
Key Features
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
More About this Item
ProCal Primary Turbidity Standards are formulated to provide instrument-specific accuracy with a safe, non toxic formulation, and offer improvements in stability over polymer and formazin alternatives, two-year shelf life. U.S. EPA-approved to meet NPDES and NPDWR compliance reporting requirements.
One 1-L bottle