HF Scientific 52400 Hach ProCal Primary Standard Kit for Portable Turbidimeter 2100P
Representative image only.
HF Scientific Mfr # 52400 – Item # EW-99900-42
Instrument specific kits and standards ensure maximum accuracy
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • NTU Value0.0, 20, 100, 800
  • Volume (mL)500
  • Volume (oz)16.9
  • Use withHach 2100P Portable Turbidimeters
  • DescriptionHach ProCal Primary Standard Kit for Portable Turbidimeter 2100P
  • Warranty1 year
1 Year Warranty
Key Features
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
More About this Item
ProCal Primary Turbidity Standards are formulated to provide instrument-specific accuracy with a safe, non toxic formulation, and offer improvements in stability over polymer and formazin alternatives, two-year shelf life. U.S. EPA-approved to meet NPDES and NPDWR compliance reporting requirements.
One 500-mL bottle of each (4 total)