High Five L973 Latex gloves, Large, 1000/cs
Representative image only.
High Five Mfr # L973 – Item # EW-86480-37
Textured finish provides enhanced grip without sacrificing tactile sensitivity
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)5
  • MaterialLatex
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • GripTextured
  • CuffBeaded
  • SterileNo
  • DescriptionLatex gloves, Large, 1000/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
E-GRIP latex gloves feature a micro-textured satin finish that enhances workers’ grip. These powder-free gloves are made using a proprietary chlorination process contain 50/µg or less of total water extractable protein per gram. Beaded cuff for easy donning. Ambidextrous.