High Five N882-10 Nitrile Gloves, Medium, 1000/cs
Representative image only.
High Five Mfr # N882-10 – Item # EW-86480-46
Formulated with skin-protecting aloe
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
PRODUCT OPTIONS
$210.00USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGreen
  • CuffBeaded
  • SterileNo
  • DescriptionNitrile Gloves, Medium, 1000/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Powder-free gloves feature a unique aloe coating on the interior glove surface for a comfortable, soothing feel against bare hands. Aloe prevents the drying and chapping that plagues frequent glove wearers.
Gloves are latex-free and contain no natural rubber proteins. Synthetic formulation offers good chemical and puncture resistance. Beaded cuff ensures easy donning. Ambidextrous.