Cole-Parmer Essentials High-purity silicone stopper, size 14 (92D)
Representative image only.
Cole-Parmer Essentials Item # EW-62994-46
Stop particle buildup
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
PRODUCT OPTIONS
$172.75USD / Pkg of 1
BUY MORE & SAVE
  • 1-5$172.75USD / Pkg of 1
  • 6-29$155.48USD / Pkg of 1
  • 30-119$146.84USD / Pkg of 1
  • 120+$138.20USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number14
  • European Size Number92D
  • Top Outside Diameter (OD) (mm)92
  • Bottom Outside Diameter (OD) (mm)79
  • Height (mm)65
  • ColorTranslucent
  • MaterialSilicone
  • DescriptionHigh-purity silicone stopper, size 14 (92D)
  • Warranty6 month
6 Month Warranty
Key Features
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
More About this Item
Translucent silicone stoppers are especially suited for procedures demanding high purity, such as tissue culture work and food science applications. Food-grade quality stoppers are nontoxic, chemically inert, and resist particle buildup. Withstand repeated sterilization, as well as temperatures from –76 to 392°F (–60 to 200°C). Peak at 428°F (220°C). European size (DIN no.) is noted on stopper. FDA compliant. Autoclavable.