Cole-Parmer Essentials High-purity silicone stopper, size 6 (31D)
Representative image only.
Cole-Parmer Essentials Item # EW-62994-22
Stop particle buildup
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
PRODUCT OPTIONS
$78.30USD / Pkg of 10
BUY MORE & SAVE
  • 1-5$78.30USD / Pkg of 10
  • 6-29$70.47USD / Pkg of 10
  • 30-119$66.56USD / Pkg of 10
  • 120+$62.64USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number6
  • European Size Number31D
  • Top Outside Diameter (OD) (mm)32
  • Bottom Outside Diameter (OD) (mm)26
  • Height (mm)30
  • ColorTranslucent
  • MaterialSilicone
  • DescriptionHigh-purity silicone stopper, size 6 (31D)
  • Warranty6 month
6 Month Warranty
Key Features
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
More About this Item
Translucent silicone stoppers are especially suited for procedures demanding high purity, such as tissue culture work and food science applications. Food-grade quality stoppers are nontoxic, chemically inert, and resist particle buildup. Withstand repeated sterilization, as well as temperatures from –76 to 392°F (–60 to 200°C). Peak at 428°F (220°C). European size (DIN no.) is noted on stopper. FDA compliant. Autoclavable.