Hirschmann 9587100 Rechargeable Battery Pack for opus® Dispenser
Representative image only.
Hirschmann Mfr # 9587100 – Item # EW-24910-50
  • Averages up to 2.5 to 3 hours of continuous operation
  • Charging time 6.5 hours
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRechargeable Battery Pack for opus Dispenser
  • Warranty1 year
1 Year Warranty
Key Features
  • Averages up to 2.5 to 3 hours of continuous operation
  • Charging time 6.5 hours
  • 1 year warranty
More About this Item
Keep your dispenser in good working order with these convenient parts and accessories.