Hollow Polypropylene (PP) Ball Bath Covers, 20 mm; 500/Pk
Representative image only.
Item # EW-06821-02
Reduce vapor and heat loss by 75% with one layer of balls
PRODUCT OPTIONS
$147.00USD / Pkg of 500
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHollow Polypropylene (PP) Ball Bath Covers, 20 mm; 500/Pk
More About this Item
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.