Honeywell 105110 Respirator Cartridges, Organic Vapor, with P100 filter, 4/box
Representative image only.
Honeywell Mfr # 105110 – Item # EW-81662-71
Screw-on cartridges install easily on respirators
SPECS & DESCRIPTION
Specifications & Description
  • RatingP100
  • NIOSH ApprovalCertain organic vapors
  • ComplianceNIOSH
  • ColorPink, Red
  • DescriptionRespirator Cartridges, Organic Vapor, with P100 filter, 4/box
More About this Item
NIOSH approved for use with organic vapors. Includes a P100 prefilter.