Honeywell 14110403 Particulate Respirators; N99, Exhalation Valve, Medium/Large, Box of 10
Representative image only.
Honeywell Mfr # 14110403 – Item # EW-81660-16
Three sizes for maximum protection, color-coded nose seal for easy size recognition
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
PRODUCT OPTIONS
$54.40USD / Box of 10
SPECS & DESCRIPTION
Specifications & Description
  • RatingN99
  • SizeMedium/large
  • ComplianceNIOSH
  • DescriptionParticulate Respirators; N99, Exhalation Valve, Medium/Large, Box of 10
Key Features
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
More About this Item
Use these respirators when working in areas containing oil-free particulate aerosols. These compact respirators are easy to use.
Approval: NIOSH
Standard models are ideal for grinding, sanding, sweeping, and other work in dusty environments. Models with exhalation valves ensure comfortable breathing even in heated environments.