Honeywell 14110439 Particulate Respirators, P100/Valve/S; 1/Bx
Representative image only.
Honeywell Mfr # 14110439 – Item # EW-81660-27
Three sizes for maximum protection, color-coded nose seal for easy size recognition
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
SPECS & DESCRIPTION
Specifications & Description
  • RatingP100
  • SizeSmall
  • ComplianceNIOSH
  • DescriptionParticulate Respirators, P100/Valve/S; 1/Bx
Key Features
  • Humidity resistant, hypoallerenic filter media for long lasting filtration
  • Boomerang nose seal securely fits various nose shapes and sizes
More About this Item
Use these respirators when working in areas containing oil-free particulate aerosols. These compact respirators are easy to use.
Approval: NIOSH
Standard models are ideal for grinding, sanding, sweeping, and other work in dusty environments. Models with exhalation valves ensure comfortable breathing even in heated environments.