Honeywell 31-80108 Laser Safety Glasses, YAG/Diode
Representative image only.
Honeywell Mfr # 31-80108 – Item # EW-86438-21
Excellent visible light transmission minimizes distortion
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorGreen
  • Lens ColorGreen
  • Laser TypeYAG, diode
  • ComplianceANSI Z136.1-1986, Z87.1-2003
  • Wavelength9 at 190-420 nm, 1 at 670 nm, 5 at 770-810 nm, 7 at 810-1100 nm, 10 at 1064nm
  • DescriptionLaser Safety Glasses, YAG/Diode
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
These lightweight, impact-resistant polymer lenses are suitable for low-powered lasers or indirect laser exposure. Wavelength, optical density, and lot number are inscribed across lens for easy identification. All eyewear comes with protective case, cleaning cloth, and safety hanging cord.
XC Series
The industry's first 9-base wraparound laser eyewear encompasses the eye while theunique Multi-Material Technology® (MMT) cushions every point of contact with the face for unparalleled comfort and impact resistance. Cushioned temples with soft, flexible nose fingers adjust to fit a variety of facial profiles. Lens and temple adjustability options further customize the fit. Detachable prescription insert is included with every pair.