This item is now discontinued. Please browse similar items
Honeywell 313000 Survivar Premier Half-Mask Respirator, Large
Representative image only.
Honeywell Mfr # 313000 – Item # EW-81661-15
Lightweight respirators feature soft sealing flange and rolled edges for added comfort
  • Excellent style and versatility
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • ComplianceNIOSH
  • ColorBlack, Blue
  • DescriptionSurvivar Premier Half-Mask Respirator, Large
Key Features
  • Excellent style and versatility
More About this Item
These lightweight respirators offer a comfortable, secure fit for excellent protection against contaminants, organic vapors, acid gases, or ammonia. Several styles are available to fit your cartridges and filters (order separately) according to the needs-order demands of your work environment.
Approval: NIOSH
Premier® Half-Mask Respirators feature a perspiration port to reduce slippage and skin irritation. Adjustable crown headstraps and suspension make adjustments easy. Soft silicone sealing flange with rolled edges provides maximum comfort. Choose an N95 particulate filter for oil-free atmospheres and P95 for protection against oil-containing aerosols. Filter base 81661-85 is required if using filter alone. Retainer cap is necessary to hold filter onto filter base or cartridge.