Honeywell 32-000460-0000 Personal Eyewash Wall Stations, 16oz single station
Representative image only.
Honeywell Mfr # 32-000460-0000-H2 – Item # EW-06794-02
Eyewash stations affix easily to any wall
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MountWall Mount
  • ComplianceANSI Z358.1-2009 as personal eyewash only
  • DescriptionPersonal Eyewash Wall Stations, 16oz single station
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Emergency eyewash stations hold one or two bottles of sterile Eyesaline® solution and affix easily to any wall with mounting screws (included). Sealed solution bottles have 36-month shelf life. Expiration date is printed on each bottle.