Honeywell B100300 Respirator Cartridges, Organic Vapors/Acid Gases, 6/box
Representative image only.
Honeywell Mfr # B100300 – Item # EW-81662-74
Screw-on cartridges install easily on respirators
SPECS & DESCRIPTION
Specifications & Description
  • RatingNone
  • NIOSH ApprovalCertain organic vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Fluoride, Hydrogen Sulfide (escape only)
  • ComplianceNIOSH
  • ColorYellow
  • DescriptionRespirator Cartridges, Organic Vapors/Acid Gases, 6/box
More About this Item
NIOSH approved for use with organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, hydrogen fluoride, and hydrogen sulfide (esc).