Honeywell B210010 Half-Mask Respirator, Small
Representative image only.
Honeywell Mfr # B210010 – Item # EW-81662-60
Silicone skirt is flexible, lightweight, and durable
  • Speaking diaphragm improves communication for added safety
SPECS & DESCRIPTION
Specifications & Description
  • SizeSmall
  • ComplianceNIOSH
  • ColorBlue
  • DescriptionHalf-Mask Respirator, Small
Key Features
  • Speaking diaphragm improves communication for added safety
More About this Item
These lightweight respirators offer a comfortable, secure fit for excellent protection against organic vapors, acid gases, contaminates, and more. Choose from several styles of cartridges and filters (order separately) according to the demands of your work environment.
Approval: NIOSH
Survivair Blue 1™ Half-Mask Respirators feature dual-flange seal for a secure seal and comfortable fit. Rear split strap makes donning easy while retainer clips hold excess strap in place. Large inhalation port lowers breathing resistance.