Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 10/XL
Representative image only.
Honeywell Mfr # 29-0910B/10XL – Item # EW-86326-67
Sturdy cut-resistant gloves simplify selection for a variety of applications
  • Color-coded marking system highlights the right gloves for your application, per EN/ANSI standards
  • Choose by level of cut resistance, protection, grip, dexterity and fit
  • One of the lightest-weight cut-protective gloves on the market
  • Touchscreen resistive and capacitive
  • Water and oil proof coating
PRODUCT OPTIONS
$18.70USD / Pair of 1
BUY MORE & SAVE
  • 1-4$18.70USD / Pair of 1
  • 5+$16.83USD / Pair of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Size10/XL
  • ColorBlack/Purple
  • LiningHPPE/Kevlar/alloy
  • ApprovalANSI A9, ISO level F
  • DescriptionHoneywell Coreshield™ 10 Gauge ANSI A9/F Glove, 10/XL
Key Features
  • Color-coded marking system highlights the right gloves for your application, per EN/ANSI standards
  • Choose by level of cut resistance, protection, grip, dexterity and fit
  • One of the lightest-weight cut-protective gloves on the market
  • Touchscreen resistive and capacitive
  • Water and oil proof coating
  • Size 10/XL
More About this Item
Providing durability and dexterity, value-packed CoreShield™ gloves from Honeywell offer optimum cut protection and user comfort. The lightweight gloves prevent hand fatigue, while a specially designed microfoam elastic coating provides outstanding oil- and dry-grip performance.
Each glove is made with softer, skin-friendly yarns that--compared to glass fiber--reduce the allergy risk to wearers. Due to this, all CoreShield™ gloves are certified STANDARD 100 by OEKO-TEX®.
CoreShield™ coatings offer 360° breathability through an open cell structure, further enhancing comfort even upon extensive wear. The washability of the glove material means little change in performance or size after five washes.