Honeywell DL4-SX1-000-000 Conductivity Electrode, K = 0.1
Representative image only.
Honeywell Mfr # DL4-SX1-000-0000 – Item # EW-56611-53
Optimize performance of your Honeywell water analysis system
  • Keep your water analysis system running properly with these accessories
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialEpoxy/graphite
  • Compatible WithHoneywell DirectLine Conductivity Transmitter, 56611-51
  • Connections3/4\" NPT(M)
  • DescriptionConductivity Electrode, K = 0.1
  • Warranty18 months
18 Month Warranty
Key Features
  • Keep your water analysis system running properly with these accessories
  • 18 month warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Honeywell water analysis system.
Conductivity electrode, epoxy body, graphite electrodes, K=0.1, 3/4''NPT mounting gland for in-line or submersion installation